missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHV-7 U69 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA567994
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The HSV Type 6 U69 protein phosphorylates the antiviral nucleoside analog ganciclovir.Spezifikation
HHV-7 U69 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Synthesized peptide derived from human GCVK_HHV7J. | |
100 μL | |
Primary | |
-20°C | |
Liquid |