Learn More
BUCHI High Performance Condenser Glass Assembly
Product Code. 17375409 Shop All Buchi Products
Change view
Click to view available options
For Use With (Equipment):
R-300, R-210, R-215
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17375409 R-300, R-210, R-215 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17375409 Supplier BUCHI Supplier No. 11066562
Verfügbar ab: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Includes a 1 liter receiving flask, the required tubing, and a ball joint clamp. For use with Rotavapor R-300, features a P+G safety coating.
Recommended in the Following Cases:
  • For reduction of solvent emissions
  • For operating at higher temperature difference (ΔT>20°C) and lower pressure values
  • For distilling low boiling solvents
  • If faster process and greater distillation rate are required
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
For Use With (Equipment) R-300, R-210, R-215
Includes Receiving Flask (1 L), Required Tubing, and a Ball Joint Clamp
Material (Coating) Plastic, Glass