Learn More
HINT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555729
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MAEEQVNRSA GLAPDCEASA TAETTVSSVG TCEAAAKSPE PKDYDSTCVF CRIAGRQDPG TELLHCENED LICFKDIKPA ATHHYLVVP Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 58%.
HINT3 Polyclonal antibody specifically detects HINT3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HINT3 | |
Polyclonal | |
Unconjugated | |
HINT3 | |
HINT3; HINT-3; HINT4; histidine triad nucleotide binding protein 3; histidine triad nucleotide-binding protein 3; HIT-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
135114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NQE9 | |
HINT3 | |
Recombinant protein corresponding to Human HINT3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only