Learn More
Epredia™ HiPur™ Methyl Alcohol
N.V.
Beschreibung
- Anhydrous
- Meets spectrophotometric grade specifications
- Final filtration is 0.2μm
Spezifikation
Methyl Alcohol |
N.V.
Methyl Alcohol |