Learn More
HIRA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562234
Additional Details : Netto-Gewicht : 0.02000kg
Description
Immunogen sequence: LKLPIPSPQR AFTLQVSSDP SMYIEVENEV TVVGGVKLSR LKCNREGKEW ETVLTSRILT AAGSCDVVCV ACEKRMLSVF STCGRRLLSP ILLPSPISTL HC Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
HIRA Polyclonal antibody specifically detects HIRA in Human samples. It is validated for Immunohistochemistry.Specifications
HIRA | |
Polyclonal | |
Unconjugated | |
HIRA | |
AA138857; D16Ertd95e; DGCR1; DiGeorge critical region gene 1; Gm15797; HIR; HIR histone cell cycle regulation defective homolog A; Hira; histone cell cycle regulation defective homolog A; histone cell cycle regulation defective homolog A (S. cerevisiae); histone cell cycle regulator; N28177; OTTMUSG00000026264; Protein HIRA; TUP1; TUP1-like enhancer of split gene 1; TUP1-like enhancer of split protein 1; TUPLE1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7290 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P54198 | |
HIRA | |
Recombinant protein corresponding to Human HIRA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only