missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2BL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548429
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with mouse based on sequence homology.
HIST1H2BL Polyclonal antibody specifically detects HIST1H2BL in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
HIST1H2BL | |
Polyclonal | |
Unconjugated | |
H2BC13 | |
dJ97D16.4; H2B histone family, member C; H2B/c; H2BFC; HIST1H2BL; histone 1, H2bl; histone cluster 1, H2bl; histone H2B type 1-L; Histone H2B.c; Histone H2Bc | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8340 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q99880 | |
H2BC13 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HIST1H2BL. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |