missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B.2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548430
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Predicted to react with mouse based on sequence homology.
Histone H2B.2 Polyclonal antibody specifically detects Histone H2B.2 in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Western Blot.Spezifikation
Histone H2B.2 | |
Polyclonal | |
Unconjugated | |
H2BC17 | |
dJ193B12.2; H2B histone family, member N; H2B.2; H2B/n; H2BFH; H2BFN; HIST1H2BO; histone 1, H2bo; histone cluster 1, H2bo; histone H2B type 1-O; Histone H2B.2; histone H2B.n; Histone H2B2; Histone H2Bn | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
8348 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P23527 | |
H2BC17 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human HIST1H2BO/HIST1H2BH. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |