missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1.3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563205
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KKAGATAGKR KASGPPVSEL ITKAVAA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 85%.
Histone H1.3 Polyclonal antibody specifically detects Histone H1.3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
Histone H1.3 | |
Polyclonal | |
Unconjugated | |
H1-3 | |
H1 histone family, member 3; H1 VAR.4; H1.3; H1D; H1f3; H1s-2; H1s-4; Hist1h1d; histone 1, family 3; histone 1, H1d; histone cluster 1, H1d; histone H1.3; Histone H1c; histone H1s-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P16402 | |
H1-3, HIST1H1D | |
Recombinant protein corresponding to Human Histone H1.3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only