missing translation for 'onlineSavingsMsg'
Learn More

Histone H2A Monoclonal Antibody (10HH2-2G12), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA3047

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 15662022

  • 471.00 € / 50 Mikroliter
Verfügbar ab: 15-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA3-047 detects Histone H2A in Human samples. MA3-047 has been succesfully used in western blot and ELISA applications. Immunogen is a synthetic peptide conjugated to ovalbumin corresponding to residues 115-124 of Human Histone H2A, which is highly conserved across multiple species.

Histone H2A Monoclonal antibody specifically detects Histone H2A in Human samples. It is validated for ELISA, Western Blot.
Spezifikation

Spezifikation

Histone H2A
Monoclonal
Conc. Not Determined
ascites with 0.05% sodium azide
P04908, P0C0S5, P0C0S8, P16104, P20671, Q96A08, Q96KK5, Q96QV6, Q99878
H2ac1, H2AC11, H2AC12, H2AC13, H2AC14, H2AC4, H2AC7, H2AC8, H2AFX, H2AX, H2AZ1, H2bc1
Synthetic peptide: VLLPKKTESH corresponding to amino acids 115-124 of human H2A.
RUO
221613, 255626, 3012, 3013, 3014, 3015, 8329, 8331, 8335, 85235, 8969
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
10HH2-2G12
Unconjugated
H2AC11
1190022L06; Acetyl-Histone H2; AW228881; dJ160A22.4; dJ86C11.1; gammaH2ax; H2A; H2A 615 antibody; H2A GL101 antibody; H2A histone; H2A histone family member; H2A histone family member X; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member A; H2A histone family, member C; H2A histone family, member E; H2A histone family, member G; H2A histone family, member M; H2A histone family, member P; H2A histone family, member Q; H2A histone family, member V; H2A histone family, member X; H2A histone family, member Z; H2A.1; H2A.1 antibody; H2A.1b; H2A.2; H2A.3; H2a.x; H2A.Z; H2A.Z1; H2a.z-1; H2A.Zl; h2a.zl1; h2a.zl2; H2A/a; H2A/c; H2A/d; H2A/E; H2A/g; H2A/m; H2A/p; H2A/q; H2A/S; H2A/X; H2A/z; H2a-291A; H2a-53; h2a-A; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; h2ad; H2ae; H2AFA; H2AFALii; H2AFC; H2AFD; H2AFE; H2AFG; H2AFI; H2AFM; H2AFN; H2AFP; H2AFQ; H2AFV; H2afx; H2afz; h2afz.L; H2AG; H2A-GL101; H2AH; h2ai; H2A-III; H2ak; H2AK5ac; H2AX; H2AX histone; H2Az;
Mouse
50 μL
Primary
Human
Antibody
IgG1 κ
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote