missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3.3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560027
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SEAYLVGLFE DTNLCAIHAK RVTIM Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Histone H3.3 Polyclonal antibody specifically detects Histone H3.3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
Histone H3.3 | |
Polyclonal | |
Unconjugated | |
H3-3A | |
9430068D06Rik; CG5825; CG5825-PA; CG5825-PC; CG5825-PD; CG8989; dH3.3A; Dmel\CG5825; Dmel_CG5825; family 3B (H3.3B); H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3.3; H3.3 histone A; H3.3 histone B; H3.3A; H3.3A/B; H3.3B; H3.4; H3.A; H3.B; H3/g; H3-3A; H3-3B; H3B; H3F3; H3F3A; H3F3A protein; H3F3B; H3FT; H3-IX; H3L-like histone; H3t; H3-X; His3.3; His3.3A; His-3.3A; His3.3A-PA; His3.3A-PC; His3.3A-PD; His3.3B; histone; Histone cluster 3; Histone H3; histone H3 class II; Histone H3.3; histone H3.3A; Histone H3.3B; histone H3.3-like protein; MGC126886; MGC126888; PH3; PP781; similar to H3 histone, family 3A; unnamed protein product | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3020, 3021 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P84243 | |
H3-3A, H3-3B | |
Recombinant protein corresponding to Human Histone H3.3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only