missing translation for 'onlineSavingsMsg'
Learn More

Histone H3, Crotonyl Lys9 Antibody (RM339), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 30027142 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mL
Unit Size:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30027142 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30027142 Supplier Novus Biologicals Supplier No. NBP326032MFV450

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, Dot Blot, Multiplex Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3
Anwendungen Western Blot, Dot Blot, Multiplex
Klassifikation Monoclonal
Klon RM339
Konjugat mFluor Violet 450 SE
Verdünnung Western Blot, Dot Blot, Multiplex Immunoassay
Zusammensetzung 50mM Sodium Borate
Gen-Alias H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
Wirtsspezies Rabbit
Immunogen This Histone H3 [Crotonyl Lys9] antibody (RM339) was raised against a crotonyl-peptide corresponding to Histone H3 [Crotonyl Lys9].
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primär oder sekundär Primary
Gen-ID (Entrez) 126961
Zielspezies Human, Vertebrates
Inhalt und Lagerung Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.