Learn More
H3K9me2S10ph Monoclonal Antibody (6HH3-2C5), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3064
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA3-064 detects dimethyl-phospho- histone H3 Lys9 pSer10 in human samples and has successfully been used in western blot and immunoprecipitation applications. The MA3-064 immunogen is a synthetic peptide conjugated to ovalbumin corresponding to residues 7-20 of Human Histone H3, which is highly conserved across multiple species.
H3K9me2S10ph Monoclonal antibody specifically detects H3K9me2S10ph in Human samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.Spezifikation
H3K9me2S10ph | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.05% sodium azide | |
P68431, Q71DI3 | |
H3C1, H3C10, H3C12, H3C14, H3C15, H3C2, H3C7 | |
Ovalbumin conjugated synthetic peptide (AR[dimethyl-K][phospho-S]TGGAPRKQLC) correspeonding to aa 7-20 of histone H3 | |
RUO | |
126961, 333932, 8350, 8356, 8357, 8358, 8968 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Immunoprecipitation, Western Blot | |
6HH3-2C5 | |
Unconjugated | |
H3C1 | |
Di histone H3/c; dimethyl H3; di-methyl H3; Dimethyl Histone H3; di-methyl histone H3; Di-Methyl-Phospho-H3 K9 S10; Di-Methyl-Phospho-H3 Lys9 Ser10; Di-Methyl-Phospho-Histone H3 K9 S10; Di-Methyl-Phospho-Histone H3 Lys9 Ser10; H3; H3 histone; H3 histone family, member A; H3 histone family, member I; H3 histone family, member K; H3 histone family, member L; H3 histone family, member M; H3 histone, family 2; H3 histone, family 3A; H3 histone, family 3B; H3 histone, family 3B (H3.3B); H3 histone, family 3C; H3 K9me2S10ph; H3.2; H3.3A; H3.3B; H3.5; H3/A; H3/b; H3/d; H3/f; H3/i; H3/j; H3/k; H3/l; H3/M; H3/n; H3/o; H3-3A; H3-3B; H3a; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3F1K; H3F2; H3F3; H3F3A; H3F3B; H3F3C; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; H3FM; H3FN; H3K9; H3K9me2; H3K9me2S10p; H3Lys9me2Ser10p; H3Lys9me2Ser10ph; H3s10; H3S10ph; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J; HIST2H3A; H | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |