missing translation for 'onlineSavingsMsg'
Learn More

HLA class I ABC Mouse anti-Human, Porcine, Clone: 5C5B7, Proteintech

Mouse Monoclonal Antibody

Marke:  Proteintech 66013-1-IG-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887505

  • 329.85 € / 150 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determits important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.
Spezifikation

Spezifikation

HLA class I ABC
5C5B7
Unconjugated
HLA-A
Liquid
HLA-A, LOC100515902
HLA class I ABC Fusion Protein Ag7488
Protein A
RUO
Antibody
Monoclonal
Human, Porcine
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
1.11 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30443
HLA A, HLA class I, HLA class I (HLA-A), HLA class I ABC, HLA-A, MHC class I antigen A*3
Mouse
IgG2b
150 μL
-20°C
Primary
100515902, 3105
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation