missing translation for 'onlineSavingsMsg'
Learn More

HLA-DPA1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 16109-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16885604

  • 399.00 € / 150 Mikroliter
Verfügbar ab: 04-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

HLA-DPA1 belongs to the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta (DPB) chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules.
Spezifikation

Spezifikation

HLA-DPA1
Polyclonal
Unconjugated
HLA-DPA1
DP(W3), DP(W4), HLA DP1A, HLA DPA1, HLA SB alpha chain, HLADP, HLADPA1, HLA-DPA1, HLASB, MHC class II DP3 alpha, MHC class II DPA1
Rabbit
Antigen Affinity Chromatography
RUO
3113
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P20036
HLA-DPA1
HLA-DPA1 Fusion Protein Ag9031
150 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote