missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Homovanillinsäure, 98+ %, Thermo Scientific Chemicals
Click to view available options
Menge:
1 g
5 g
25 g
Descrizione
Fluorimetric reagent and major catecholamine metaboliteHomovanillic acid is used as a reagent in the determination of oxidative enzymes. It is associated with dopamine levels in the brain. Its presence supports a diagnosis of neuroblastoma and malignant pheochromocytoma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Löslichkeit
Löslich in Wasser, Dimethylsulfoxid, Ethanol, Methanol und Ethylacetat.
Hinweise
Lichtempfindlich. Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
Löslich in Wasser, Dimethylsulfoxid, Ethanol, Methanol und Ethylacetat.
Hinweise
Lichtempfindlich. Kühl lagern. Nicht kompatibel mit starken Oxidationsmitteln.
Specifica
Chemischer Name oder Material | Homovanillic acid |
Schmelzpunkt | 140°C to 145°C |
Menge | 1 g |
Beilstein | 2213447 |
Merck Index | 14,4740 |
Löslichkeitsinformationen | Soluble in water,dimethyl sulfoxide,ethanol,methanol and ethyl acetate. |
Formelmasse | 182.18 |
Reinheit (%) | ≥98% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'