missing translation for 'onlineSavingsMsg'
Learn More

HRP-1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

N.V.

Spezifikation

Antigen HRP-1/HDGFL1
Immunogen E. coli-derived recombinant human HRP-1 Ser2-Leu283 Accession # NP_032258
Gen-ID (Entrez) 154150
Gen-Zugriffsnummer NP_032258
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

HRP-1/HDGFL1 Polyclonal specifically detects HRP-1/HDGFL1 in Mouse samples. It is validated for Western Blot.
Spezifikation

Spezifikation

HRP-1/HDGFL1
154150
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects mouse HRP-1 in direct ELISAs and Western blots.
Unconjugated
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
E. coli-derived recombinant human HRP-1 Ser2-Leu283 Accession # NP_032258
NP_032258
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only