Learn More
HS2ST1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560228
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NQIQKLEESR SKLERAIARH EVREIEQRHT MDGPRQDATL DEEEDMVIIY NRVPKTASTS FTNIAYDLCA KNKYHVLH Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.
HS2ST1 Polyclonal antibody specifically detects HS2ST1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
HS2ST1 | |
Polyclonal | |
Unconjugated | |
HS2ST1 | |
2OST; 2-OST; 2-O-sulfotransferase; AW214369; dJ604K5.2; heparan sulfate 2-O-sulfotransferase 1; Hs2st; Hs2st1; KIAA0448; mKIAA0448 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9653 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7LGA3 | |
HS2ST1 | |
Recombinant protein corresponding to Human HS2ST1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only