missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564210
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ALDPSQSFKV YATLRDLKTQ GRLWEAARAL ACPQG Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 77%.
HSD17B1 Polyclonal antibody specifically detects HSD17B1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
HSD17B1 | |
Polyclonal | |
Unconjugated | |
HSD17B1 | |
17-beta-dehydrogenase 1; 17beta-HSD; 17-beta-HSD 1; 17-beta-hydroxysteroid dehydrogenase type 1; 17beta-hydroxysteroid dehydrogenase type 1; 17BHD1; 17HSDB1; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E17KSR; E2DH; Edh17b1; EDH17B2; EDHB17; Estradiol 17-beta-dehydrogenase 1; estradiol 17-beta-dehydrogenase-1; HSD17; HSD17B1; Hsd17ba; hydroxysteroid (17-beta) dehydrogenase 1; hydroxysteroid (17-beta) dehydrogenase 1 isoform; hydroxysteroid 17-beta dehydrogenase 1; Hydroxysteroid dehydrogenase 17 beta, type 1; LOW QUALITY PROTEIN: estradiol 17-beta-dehydrogenase 1; placental 17-beta-hydroxysteroid dehydrogenase; SDR28C1; Short chain dehydrogenase/reductase family 28C member 1; short chain dehydrogenase/reductase family 28CE, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3292 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P14061 | |
HSD17B1 | |
Recombinant protein corresponding to Human HSD17B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only