Learn More
HSD17B2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562157
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LVTGGDCGLG HALCKYLDEL GFTVFAGVLN ENGPGAEELR RTCSPRLSVL QMDITKPVQI KDAYSKVAAM LQDRGLWAVI NNAGVLGFPT DGELLLMTDY KQCMAVNFFG TV Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 73%.
HSD17B2 Polyclonal antibody specifically detects HSD17B2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
HSD17B2 | |
Polyclonal | |
Unconjugated | |
HSD17B2 | |
1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; AI194836; AI194967; AI255511; E2DH; EDH17B2; estradiol 17-beta-dehydrogenase 2; HSD17; HSD17B2; hydroxysteroid (17-beta) dehydrogenase 2; hydroxysteroid 17-beta dehydrogenase 2; microsomal 17-beta-hydroxysteroid dehydrogenase; SDR9C2; short chain dehydrogenase/reductase family 9C member 2; short chain dehydrogenase/reductase family 9C, member 2; tes; testosterone 17-beta-dehydrogenase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3294 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P37059 | |
HSD17B2 | |
Recombinant protein corresponding to Human HSD17B2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only