missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP701L Monoclonal Antibody (3G10), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA524381
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous HSP701L / Heat Shock Protein 70 1-like proteins.
HSP701L Monoclonal antibody specifically detects HSP701L in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
HSP701L | |
Monoclonal | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P16627, P34931, P55063 | |
Hspa1l | |
Recombinant Protein of Human HSP70 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
3G10 | |
Unconjugated | |
Hspa1l | |
heat shock 10kDa protein 1-like; heat shock 70 kDa protein 1-Hom; heat shock 70 kDa protein 1L; Heat shock 70 kDa protein 1-like; Heat shock 70 kDa protein 3; heat shock 70 kDa-like protein 1; heat shock 70kD protein-like 1; heat shock 70kDa protein 1-like; Heat shock protein; heat shock protein 1-like; Heat shock protein 70-1l; heat shock protein cognate 70, testis; heat shock protein family A (Hsp70) member 1 like; heat shock-related protein hsc70t; Hsc70t; HSP70.3; HSP70-1L; Hsp70-3; HSP70-HOM; HSP70T; Hspa1l; hum70t; inducible heat shock protein 70; Msh5; Spermatid-specific heat shock protein 70 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
15482, 24963, 3305 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |