Learn More
HSPA12B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553354
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LAVPEMGLQG LYIGSSPERS PVPSPPGSPR TQESCGIAPL TPSQSPKPEV RAPQQASFSV VVAIDFGTTS SGYAFSFASD PEAIHMMRKW EGG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
HSPA12B Polyclonal antibody specifically detects HSPA12B in Human samples. It is validated for Immunohistochemistry.Spezifikation
HSPA12B | |
Polyclonal | |
Unconjugated | |
HSPA12B | |
2700081N06Rik; C20orf60; Heat shock 70 kDa protein 12B; Heat shock protein; heat shock protein 12B; heat shock protein 70kDa 12B; heat shock protein family A (Hsp70) member 12B; HSP; HSPA12B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116835 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
Lot-Specific | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96MM6 | |
HSPA12B | |
Recombinant protein corresponding to Human HSPA12B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only