missing translation for 'onlineSavingsMsg'
Learn More

HSPB11 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA556362

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15774271

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: IDLCLSSEGS EVILATSSDE KHPPENIIDG NPETFWTTTG MFPQEFIICF HKHVRIERLV IQSYFVQT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

HSPB11 Polyclonal antibody specifically detects HSPB11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

HSPB11
Polyclonal
Unconjugated
Hspb11
2900042B11Rik; C1orf41; Heat shock protein; Heat shock protein beta-11; heat shock protein family B (small) member 11; heat shock protein family B (small), member 11; HSP; Hspb11; Hspb11 heat shock protein family B (small), member 11; HSPC034; HSPCO34; IFT25; intraflagellar transport 25 homolog; intraflagellar transport protein 25 homolog; Placental protein 25; placental protein 25 homolog; PP25
Rabbit
Antigen affinity chromatography
RUO
51668
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9Y547
Hspb11
Recombinant protein corresponding to Human HSPB11
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only