Learn More
ACROBiosystems Human Activin RIIB / ACVR2B Protein, His Tag
314.52 € - 1342.71 €
Spezifikation
Zugriffsnummer | NP_001097.2 |
---|---|
Molekulargewicht | 14.5 kDa |
Quelle | Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1). |
Immunogen | Ser 19 - Thr 137 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15895877
|
ACROBiosystems
ACB-H5226-200UG |
200 μg |
314.52 €
200 Mikrogramm |
Verfügbar ab: 30-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15885877
|
ACROBiosystems
ACB-H5226-1MG |
1 mg |
1342.71 €
1 Milligramm |
Verfügbar ab: 30-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
Activin receptor type-2B (ACVR2B) is also known as ActR-IIB and MGC116908, ACVR2B is an activin type 2 receptor. Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I (I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Type II receptors are considered to be constitutively active kinases. This gene encodes activin A type IIB receptor, which displays a 3- to 4-fold higher affinity for the ligand than activin A type II receptor. Defects in ACVR2B are the cause of visceral heterotaxy autosomal type 4 (HTX4).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.5 kDa. The protein migrates as 22-44 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001097.2 | |
Human Activin RIIB, His Tag (ACB-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 19 - Thr 137 (Accession # Q13705-1). | |
-20°C | |
Activin RIIB | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Other Recombinant Proteins |
14.5 kDa | |
Ser 19 - Thr 137 | |
ACVR2B,ACTRIIB,MGC116908 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |