Learn More
Abnova™ Human ADCY3 Partial ORF (NP_004027, 1035 a.a. - 1144 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000109-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes adenylyl cyclase 3 which is a membrane-associated enzyme and catalyzes the formation of the secondary messenger cyclic adenosine monophosphate (cAMP). This protein appears to be widely expressed in various human tissues and may be involved in a number of physiological and pathophysiological metabolic processes. [provided by RefSeq]
Sequence: FNNFMLRIGMNKGGVLAGVIGARKPHYDIWGNTVNVASRMESTGVMGNIQVVEETQVILREYGFRFVRRGPIFVKGKGELLTFFLKGRDKLATFPNGPSVTLPHQVVDNSSpezifikation
NP_004027 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FNNFMLRIGMNKGGVLAGVIGARKPHYDIWGNTVNVASRMESTGVMGNIQVVEETQVILREYGFRFVRRGPIFVKGKGELLTFFLKGRDKLATFPNGPSVTLPHQVVDNS | |
RUO | |
ADCY3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
109 | |
ADCY3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AC3/KIAA0511 | |
ADCY3 | |
Recombinant | |
wheat germ expression system |