Learn More
Abnova™ Human AGPAT3 Full-length ORF (AAH04219.1, 1 a.a. - 63 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00056894-P02.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is an acyltransferase that converts lysophosphatidic acid into phosphatidic acid, which is the second step in the de novo phospholipid biosynthetic pathway. The encoded protein may be an integral membrane protein. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq]
Sequence: MGSPAHLPKDAIAVFIVFPRVRHWLSGTRRQCTRRAPYVRCFKVPVLRKRSLVEEMSAEQITKSpezifikation
AAH04219.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
AGPAT3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
56894 | |
AGPAT3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LPAAT-GAMMA1/MGC4604 | |
AGPAT3 | |
Recombinant | |
wheat germ expression system |