Learn More
Abnova™ Human ANAPC4 Partial ORF (AAH59383, 651 a.a. - 750 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00029945-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits. The composition of the APC is highly conserved in organisms from yeast to humans. The exact function of this gene product is not known. [provided by RefSeq]
Sequence: VVLKDTVGREGRDRLLVQLPLSLVYNSEDSAEYQFTGTYSTRLDEQCSAIPTRTMHFEKHWRLLESMKAQYVAGNGFRKVSCVLSSNLRHVRVFEMDIDDSpezifikation
AAH59383 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VVLKDTVGREGRDRLLVQLPLSLVYNSEDSAEYQFTGTYSTRLDEQCSAIPTRTMHFEKHWRLLESMKAQYVAGNGFRKVSCVLSSNLRHVRVFEMDIDD | |
RUO | |
ANAPC4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29945 | |
ANAPC4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
APC4 | |
ANAPC4 | |
Recombinant | |
wheat germ expression system |