Learn More
Abnova™ Human ANXA2 Partial ORF (AAH09564.1, 186 a.a. - 268 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000302-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. This protein functions as an autocrine factor which heightens osteoclast formation and bone resorption. This gene has three pseudogenes located on chromosomes 4, 9 and 10, respectively. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: IDYELIDQDARDLYDAGVKRKGTDVPKWISIMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPLSpezifikation
AAH09564.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.76kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IDYELIDQDARDLYDAGVKRKGTDVPKWISIMTERSVPHLQKVFDRYKSYSPYDMLESIRKEVKGDLENAFLNLVQCIQNKPL | |
RUO | |
ANXA2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
302 | |
ANXA2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANX2/ANX2L4/CAL1H/LIP2/LPC2/LPC2D/P36/PAP-IV | |
ANXA2 | |
Recombinant | |
wheat germ expression system |