missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Apo-SAA1 Recombinant Protein, PeproTech®

Artikelnummer. 17882443
Change view
Click to view available options
Menge:
10 μg
100 μg
2 x 500 μg
250 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
10 Mikrogramm
100 Mikrogramm
250 Mikrogramm
50 Mikrogramm
500 Mikrogramm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17882443 10 μg 10 Mikrogramm
17802203 100 μg 100 Mikrogramm
17892393 2 x 500 μg 1 Milligramm
17822343 250 μg 250 Mikrogramm
17872443 500 μg 500 Mikrogramm
17822683 50 μg 50 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17882443 Lieferant Gibco™ Lieferanten-Nr. 3005310UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

300-53-1MG will be provided as 2 x 500 μg (300-53-500UG). Recombinant Human Apo-SAA1 is an 11.7 kDa protein containing 105 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The serum amyloid A family comprises a number of differentially expressed apolipoproteins, acute-phase SAA1 and SAA2, the former being the major component in plasma and constitutive SAAs. Although the liver is the primary site of synthesis of both SAA types extrahepatic production has been reported. The in vivo concentrations increase by as much as 1000 fold during inflammation. Several studies have expressed its importance in the diagnosis and monitoring of various diseases. Pathological SAA values are often detected in association with normal CRP concentrations; SAA rises earlier and more sharply than CRP. Recently, a broader view of SAA expression and function has been emerging.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer P0DJI8
Zur Verwendung mit (Anwendung) Control, Bioactivity
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 6288
Molekulargewicht 11.7 kDa
Name Human Apo-SAA1
Reinigungsverfahren Purified
Menge 10 μg
Quelle E. coli
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/μg
Gen-Alias amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4
Gebräuchliche Bezeichnung SAA
Gensymbol SAA1
Biologische Aktivität Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz MRSFFSFLGE AFDGARDMWR AYSDMREANY IGSDKYFHAR GNYDAAKRGP GGVWAAEAIS DARENIQRFF GHGAEDSLAD QAANEWGRSG KDPNHFRPAG LPEKY
Inhalt und Lagerung -20°C
Aktivität Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml.
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.