Learn More
ACROBiosystems Human B7-H2 / ICOSLG Protein, His Tag
Marke:  ACROBiosystems B72-H5221-200UG
Weitere Details : Netto-Gewicht : 0.00001kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15895947
  • 307.93 € / 200 Mikrogramm
Verfügbar ab: 05-01-2023
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
ICOS ligand (ICOSLG) is also known as B7 homolog 2 (B7-H2), B7-related protein 1 (B7RP-1) and CD antigen CD275, which belongs to the immunoglobulin superfamily and BTN/MOG family. ICOSLG contains one Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. Isoform 1 is widely expressed, while isoform 2 is detected only in lymph nodes, leukocytes and spleen. B7-H2 is ligand for the T-cell-specific cell surface receptor ICOS. B7-H2 acts as a costimulatory signal for T-cell proliferation and cytokine secretion and induces also B-cell proliferation and differentiation into plasma cells. B7-H2 could play an important role in mediating local tissue responses to inflammatory conditions, as well as in modulating the secondary immune response by co-stimulating memory T-cell function.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 27.8 kDa. The protein migrates as 45-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_056074
27.8 kDa
200 µg
Asp 19 - Ser 258
ICOSLG,B7-H2,B7H2,B7RP-1,B7RP1,CD275,GL50,ICOS-L,ICOSL,LICOS,ICOS ligand
Unconjugated
PBS, pH7.4
HEK293
Human
Powder
>95% as determined by SDS-PAGE
Human B7-H2, His Tag (B72-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 19 - Ser 258 (Accession # NP_056074).
-20°C
B7-H2
His Tag
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins