Learn More
Abnova™ Human BAT4 Partial ORF (NP_149417, 257 a.a. - 356 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00007918-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
A cluster of genes, BAT1-BAT5, has been localized in the vicinity of the genes for TNF alpha and TNF beta. These genes are all within the human major histocompatibility complex class III region. The protein encoded by this gene is thought to be involved in some aspects of immunity. [provided by RefSeq]
Sequence: SPGFKLLLRGGWEPGMGLGPRGEGRANPIPTVLKRDQEGLGYRSAPQPRVTHFPAWDTRAVAGRERPPRVATLSWREERRREEKDRAWERDLRTYMNLEFSpezifikation
NP_149417 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SPGFKLLLRGGWEPGMGLGPRGEGRANPIPTVLKRDQEGLGYRSAPQPRVTHFPAWDTRAVAGRERPPRVATLSWREERRREEKDRAWERDLRTYMNLEF | |
RUO | |
BAT4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7918 | |
BAT4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANKRD59/D6S54E/G5/GPANK1/GPATCH10 | |
BAT4 | |
Recombinant | |
wheat germ expression system |