missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BIK Partial ORF (AAH01599, 37 a.a. - 136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifica
Zugriffsnummer | AAH01599 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 638 |
Molekulargewicht | 36.63kDa |
Codice del prodotto | Marca | Menge | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Menge | Prezzo | Quantità e disponibilità | |||||
16172241
|
Abnova™
H00000638-Q01.25UG |
25 ug |
508.00 €
25 Mikrogramm |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16162241
|
Abnova™
H00000638-Q01.10UG |
10 ug |
335.00 €
10 Mikrogramm |
Verfügbar ab: 05-06-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Descrizione
The protein encoded by this gene is known to interact with cellular and viral survival-promoting proteins, such as BCL2 and the Epstein-Barr virus in order to enhance programed cell death. Because its activity is suppressed in the presence of survival-promoting proteins, this protein is suggested as a likely target for antiapoptotic proteins. This protein shares a critical BH3 domain with other death-promoting proteins, BAX and BAK. [provided by RefSeq]
Sequence: EDLDPMEDFDSLECMEGSDALALRLACIGDEMDVSLRAPRLAQLSEVAMHSLGLAFIYDQTEDIRDVLRSFMDGFTTLKENIMRFWRSPNPGSWVSCEQVSpecifica
AAH01599 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIP1/BP4/NBK | |
BIK | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
638 | |
BIK (Human) Recombinant Protein (Q01) | |
EDLDPMEDFDSLECMEGSDALALRLACIGDEMDVSLRAPRLAQLSEVAMHSLGLAFIYDQTEDIRDVLRSFMDGFTTLKENIMRFWRSPNPGSWVSCEQV | |
RUO | |
BIK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |