missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human BTLA (31-150) Protein, Fc Tag

Marke:  ACROBiosystems BTA-H5256-100UG

Weitere Details : Netto-Gewicht : 0.00001kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 15846067

  • 353.08 € / 100 Mikrogramm

Voraussichtliches Versanddatum: 22-11-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 40.4 kDa. The protein migrates as 46-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAP44003.1
HEK293
Human
40.4 kDa
100 µg
Lys 31 - Ser 150
BTLA,CD272
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>92% as determined by SDS-PAGE
Human BTLA, Fc Tag (BTA-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Ser 150 (Accession # AAP44003.1).
-20°C
BTLA
Fc Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate