missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C9orf19 Partial ORF (NP_071738.1, 2 a.a. - 86 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00152007-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Sequence: GKSASKQFHNEVLKAHNEYRQKHGVPPLKLCKNLNREAQQYSEALASTRILKHSPESSRGQCGENLAWASYDQTGKEVADRWYSESpezifikation
NP_071738.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.09kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GKSASKQFHNEVLKAHNEYRQKHGVPPLKLCKNLNREAQQYSEALASTRILKHSPESSRGQCGENLAWASYDQTGKEVADRWYSE | |
RUO | |
GLIPR2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
152007 | |
C9orf19 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C9orf19/GAPR-1 | |
GLIPR2 | |
Recombinant | |
wheat germ expression system |