missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CAPS2 Partial ORF (NP_115995.1, 285 a.a. - 382 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00084698-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).[supplied by OMIM]
Sequence: YRKSYVRKAFMKLDFNKSGSVPIINIRKCYCAKKHSQVISGHSTEEEIKSSFLETLKVACSKSDEVSYGEFEDYYEGLSIGIVDDEDFVNILRTPWGISpezifikation
NP_115995.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
YRKSYVRKAFMKLDFNKSGSVPIINIRKCYCAKKHSQVISGHSTEEEIKSSFLETLKVACSKSDEVSYGEFEDYYEGLSIGIVDDEDFVNILRTPWGI | |
RUO | |
CAPS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
84698 | |
CAPS2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ34520/UG0636c06 | |
CAPS2 | |
Recombinant | |
wheat germ expression system |