missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CASC4 Full-length ORF (NP_612432.2, 1 a.a. - 436 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00113201-P01.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166983

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

The increased expression level of this gene is associated with HER-2/neu proto-oncogene overexpression. Amplification and resulting overexpression of this proto-oncogene are found in approximately 30% of human breast and 20% of human ovarian cancers. Further characterization of the product of this gene may yield insight into the fundamental biology and pathogenetic effects of HER-2/neu overexpression in human breast and ovarian cancer cells. Alternatively spliced variants encoding different isoforms have been identified for this gene. [provided by RefSeq]

Sequence: MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVL
Spezifikation

Spezifikation

NP_612432.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
75.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp459F1927/H63/MGC74708
CASC4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
113201
CASC4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVGFGANRRAGRLPSLVLVVLLVVIVVLAFNYWSISSRHVLLQEEVAELQGQVQRTEVARGRLEKRNSDLLLLVDTHKKQIDQKEADYGRLSSRLQAREGLGKRCEDDKVKLQNNISYQMADIHHLKEQLAELRQEFLRQEDQLQDYRKNNTYLVKRLEYESFQCGQQMKELRAQHEENIKKLADQFLEEQKQETQKIQSNDGKELDINNQVVPKNIPKVAENVADKNEEPSSNHIPHGKEQIKRGGDAGMPGIEENDLAKVDDLPPALRKPPISVSQHESHQAISHLPTGQPLSPNMPPDSHINHNGNPGTSKQNPSSPLQRLIPGSNLDSEPRIQTDILKQATKDRVSDFHKLKQSRFFDENESPVDPQHGSKLADYNGDDGNVGEYEADKQAELAYNEEEDGDGGEEDVQDDEERELQMDPADYGKQHFNDVL
RUO
CASC4
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote