missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL2/MCP-1 DuoSet, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human CCL2/MCP-1

Marke:  R&D Systems™ DY279

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413475

  • 666.85 € / 15 Stück


Voraussichtlicher Liefertermin 02-05-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation

Spezifikation

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
6347
15 Plates
Human
15.60 - 1000pg/mL
Videos