missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL25 (TECK) Recombinant Protein, PeproTech®

Artikelnummer. 17812853
Change view
Click to view available options
Menge:
1 mg
100 μg
20 μg
250 μg
5 μg
50 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
5 Mikrogramm
50 Mikrogramm
500 Mikrogramm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17812853 250 μg 250 Mikrogramm
17892953 100 μg 100 Mikrogramm
17852753 1 mg 1 Milligramm
17882653 20 μg 20 Mikrogramm
17882553 500 μg 500 Mikrogramm
17832883 50 μg 50 Mikrogramm
17882143 5 μg 5 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17812853 Lieferant Gibco™ Lieferanten-Nr. 30045250UG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Recombinant Human TECK is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Chemokines are likely to play an important role in regulating the trafficking of developing T cells within the thymus. Chemokine C-C thymus expressed chemokine (TECK), also designated chemokine ligand 25 (CCL25), small induc-ible cytokine A25, chemokine b-15 or CK b-15, is expressed predominantly in thymic dendritic cells, thymic epithelial cells and in the small intestine. TECK, a CCR9 ligand, has suppressive activity against immature subsets of myeloid progenitors which have been stimulated to proliferate by multiple growth factors. TECK delivers signals through CCR9, which is important for the nav-igation of developing thymocytes. Bone marrow pre-pro-B cells and cells capable of generating pro-B colonies in the presence of interleukin-7 and Flt-3 ligand migrate to TECK, a response lost in later stages of B cell development.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer O15444
Zur Verwendung mit (Anwendung) Control, Western Blot Control, ELISA Standard, Binding Assay, Bioactivity
Zusammensetzung Protein with no preservative
Gen-ID (Entrez) 6370
Molekulargewicht 14.2 kDa
Name Human CCL25 (TECK)
Reinigungsverfahren Purified
Menge 250 μg
Quelle E. coli
Lagerungsbedingungen -20°C
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/μg
Gen-Alias A130072A22Rik; AI852536; C-C motif chemokine 25; C-C motif chemokine ligand 25; CCL25; chemokine (C-C motif) ligand 25; chemokine TECK; Ck beta-15; CKb15; Scya25; SCYA25TECKvar; small inducible chemokine 25; small inducible cytokine A25; small inducible cytokine subfamily A (Cys-Cys), member 25; small-inducible cytokine A25; Teck; TECKMGC150327; TECKvar; thymus expressed chemokine; thymus-expressed chemokine
Gebräuchliche Bezeichnung TECK
Gensymbol CCL25
Biologische Aktivität Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Produkttyp Protein
Konjugat Unconjugated
Rekombinant Recombinant
Sequenz QGVFEDCCLA YHYPIGWAVL RRAWTYRIQE VSGSCNLPAA IFYLPKRHRK VCGNPKSREV QRAMKLLDAR NKVFAKLHHN MQTFQAGPHA VKKLSSGNSK LSSSKFSNPI SSSKRNVSLL ISANSGL
Inhalt und Lagerung -20°C
Aktivität Determined by its ability to bind CCR9 receptor expressed on the surface of MOLT4 cells.
Expressionssystem E. coli
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.