missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CCL3/MIP-1 alpha DuoSet, R&D Systems™

Human CCL3/MIP-1 alpha DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

N.V.

Spezifikation

Assay-Bereich 7.81 to 500 pg/mL
Kreuzreaktivität <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Gen-ID (Entrez) 6348
Produkttyp ELISA Kit
Format 15 96-well Plates
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

The Human CCL3/MIP-1 alpha DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CCL3/MIP-1 alpha.
Spezifikation

Spezifikation

7.81 to 500 pg/mL
6348
15 96-well Plates
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Kit
Biotin
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen