Learn More
R&D Systems™ Human CCL3/MIP-1 alpha DuoSet, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure natural and recombinant Human CCL3/MIP-1 alpha
Marke:  R&D Systems™ DY270
Weitere Details : Netto-Gewicht : 0.01000kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13413455
  • 666.85 € / 15 Stück
Voraussichtlicher Liefertermin 04-06-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Spezifikation
Spezifikation
ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
6348
1 kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
15 96-well plates
ELISA kit
Human
7.81 - 500pg/mL
Videos