missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CD160 Full-length ORF (NP_008984.1, 1 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00011126-P02.10ug

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16166202

  • 335.00 € / 10 Mikrogramm
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

CD160 is an 27 kDa glycoprotein which was initially identified with the monoclonal antibody BY55. Its expression is tightly associated with peripheral blood NK cells and CD8 T lymphocytes with cytolytic effector activity. The cDNA sequence of CD160 predicts a cysteine-rich, glycosylphosphatidylinositol-anchored protein of 181 amino acids with a single Ig-like domain weakly homologous to KIR2DL4 molecule. CD160 is expressed at the cell surface as a tightly disulfide-linked multimer. RNA blot analysis revealed CD160 mRNAs of 1.5 and 1.6 kb whose expression was highly restricted to circulating NK and T cells, spleen and small intestine. Within NK cells CD160 is expressed by CD56dimCD16+ cells whereas among circulating T cells its expression is mainly restricted to TCRgd bearing cells and to TCRab+CD8brightCD95+CD56+CD28-CD27-cells. In tissues, CD160 is expressed on all intestinal intraepithelial lymphocytes. CD160 shows a broad specificity for binding to both classical and nonclassical MHC class I molecules. [provided by RefSeq]

Sequence: MLLEPGRGCCALAILLAIVDIQSGGCINITSSASQEGTRLNLICTVWHKKEEAEGFVVFLCKDRSGDCSPETSLKQLRLKRDPGIDGVGEISSQLMFTISQVTPLHSGTYQCCARSQKSGIRLQGHFFSILFTETGNYTVTGLKQRQHLEFSHNEGTLSSGFLQEKVWVMLVTSLVALQAL
Spezifikation

Spezifikation

NP_008984.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
46.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BY55/FLJ46513/NK1/NK28
CD160
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
11126
CD160 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLLEPGRGCCALAILLAIVDIQSGGCINITSSASQEGTRLNLICTVWHKKEEAEGFVVFLCKDRSGDCSPETSLKQLRLKRDPGIDGVGEISSQLMFTISQVTPLHSGTYQCCARSQKSGIRLQGHFFSILFTETGNYTVTGLKQRQHLEFSHNEGTLSSGFLQEKVWVMLVTSLVALQAL
RUO
CD160
Wheat Germ (in vitro)
GST
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote