Learn More
ACROBiosystems Human CD2 / SRBC Protein, His Tag
423.38 € - 2951.55 €
Spezifikation
Zugriffsnummer | AAH33583 |
---|---|
Molekulargewicht | 22.3 kDa |
Quelle | Human CD2, His Tag (CD2-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583). |
Immunogen | Lys 25 - Asp 209 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15806157
|
ACROBiosystems
CD2-H5226-50UG |
50 μg |
423.38 €
50 Mikrogramm |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15896147
|
ACROBiosystems
CD2-H5226-1MG |
1 mg |
2951.55 €
1 Milligramm |
Verfügbar ab: 29-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
T-cell surface antigen CD2 is also known as Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5 and SRBC, is a single-pass type I membrane protein found on the surface of T cells and natural killer (NK) cells. CD2 is a member of the immunoglobulin superfamily. CD2 / SRBC contains 1 Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain. CD2 / SRBC interacts with other adhesion molecules, such as lymphocyte function-associated antigen-3 (LFA-3 / CD58) in humans, or CD48 in rodents, which are expressed on the surfaces of other cells. In addition to its adhesive properties, CD2 also acts as a co-stimulatory molecule on T and NK cells. CD2 is a specific marker for T cells and NK cells, and can therefore be used in immunohistochemistry to identify the presence of such cells in tissue sections.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH33583 | |
Human CD2, His Tag (CD2-H5226) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accession # AAH33583). | |
-20°C | |
CD2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
22.3 kDa | |
Lys 25 - Asp 209 | |
CD2,SRBC,LFA-2,T11 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |