Learn More
ACROBiosystems Human CD200 / OX-2 Protein, His Tag
302.42 € - 1935.44 €
Spezifikation
Zugriffsnummer | AAH22522 |
---|---|
Molekulargewicht | 23.3 kDa |
Quelle | Human CD200, His Tag (O x 2-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522). |
Immunogen | Gln 31 - Gly 232 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15887407
|
ACROBiosystems
OX2-H5228-100UG |
100 μg |
302.42 €
100 Mikrogramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15897407
|
ACROBiosystems
OX2-H5228-1MG |
1 mg |
1935.44 €
1 Milligramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD200 is also known as OX-2 membrane glycoprotein (OX-2), is a type-1 membrane glycoprotein, which contains two immunoglobulin domains (1 Ig-like C2-type (immunoglobulin-like) domain and Ig-like V-type (immunoglobulin-like) domain), and thus belongs to the immunoglobulin superfamily. CD200 / OX-2 is widely expressed in multiple cell types. CD200 interacts with a structurally related receptor (CD200R) expressed mainly on myeloid cells and is involved in regulation of macrophage and mast cell function. OX-2 / CD200 and CD200R associate via their respective N-terminal Ig-like domains. CD200 also plays an important role in prevention of graft rejection, autoimmune diseases and spontaneous abortion.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.3 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH22522 | |
Human CD200, His Tag (O x 2-H5228) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522). | |
-20°C | |
CD200 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
23.3 kDa | |
Gln 31 - Gly 232 | |
CD200,MOX1,MOX2,MRC,OX-2,My033 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |