Learn More
R&D Systems™ Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit
Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
N.V.
Spezifikation
Assay-Bereich 62.50 to 4000 pg/mL
Assay-Empfindlichkeit: 10.1 pg/mL
Probentyp Cell Culture Supernatant, Serum, Platelet-poor Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Serum
Mehr Spezifikationen ansehen
Beschreibung
Beschreibung
The Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CD40 Ligand/TNFSF5 in cell culture supernates (100 ul), serum (20 ul), platelet-poor edta plasma (100 ul), platelet-poor heparin plasma (100 ul).
Spezifikation
Spezifikation
62.50 to 4000 pg/mL
10.1 pg/mL
Cell Culture Supernatant, Serum, Platelet-poor Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Serum