missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human CD40 Ligand/TNFSF5 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human CD40 Ligand in cell culture supernates, serum, and plasma.

Marke:  R&D Systems™ DCDL40

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412615

  • 774.36 € / 96 Tests


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Quantikine Human CD40 Ligand Immunoassay is a 4.5 hour solid-phase ELISA designed to measure human CD40 Ligand in cell culture supernates, serum, and plasma. It contains E. coli-expressed recombinant human CD40 Ligand and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human CD40 Ligand showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring human CD40 Ligand.
Spezifikation

Spezifikation

62.50 - 4000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
959
1 Kit
Cell Culture Supernates (100uL), Serum (20uL), Platelet-poor EDTA Plasma (100uL), Platelet-poor Heparin Plasma (100uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
1 Plate
10.1pg/mL
Cell Culture Supernates (100μL), Serum (20μL), Platelet-poor EDTA Plasma (100μL), Platelet-poor Heparin Plasma (100μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos