Learn More
Abcam Human CDC42 knockout HEK-293T cell line
Product Code. 32141217 Shop All Abcam Products
Change view
Click to view available options
Quantity:
1,000,000 Cells
2 x 1,000,000 Cells
Unit Size:
Each
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32141217 1,000,000 Cells Each
32139934 2 x 1,000,000 Cells Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 32141217 Supplier Abcam Supplier No. ab2665221000000Cells_
 Restricted Item
Verfügbar ab: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
CDC42 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 2 bp insertion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockou...
CDC42 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 2 bp insertion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 1,000,000 Cells