Learn More
Abnova™ Human CDH18 Partial ORF (NP_004925, 467 a.a. - 576 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00001016-Q01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a type II classical cadherin from the cadherin superfamily of integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. This particular cadherin is expressed specifically in the central nervous system and is putatively involved in synaptic adhesion, axon outgrowth and guidance. [provided by RefSeq]
Sequence: DLLSHVTVGIRVLDVNDNPPELAREYDIIVCENSKPGQVIHTISATDKDDFANGPRFNFFLDERLPVNPNFTLKDNEDNTASILTRRRRFSRTVQDVYYLPIMISDGGIPSpezifikation
NP_004925 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DLLSHVTVGIRVLDVNDNPPELAREYDIIVCENSKPGQVIHTISATDKDDFANGPRFNFFLDERLPVNPNFTLKDNEDNTASILTRRRRFSRTVQDVYYLPIMISDGGIP | |
RUO | |
CDH18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1016 | |
CDH18 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDH14/CDH14L/CDH24/EY-CADHERIN | |
CDH18 | |
Recombinant | |
wheat germ expression system |