missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Clusterin Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid-phase ELISA designed to measure human Clusterin in cell culture supernates, serum, plasma, urine, and saliva.

Marke:  R&D Systems™ SCLU00

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412696

  • 4428.58 € / 1 Set


Voraussichtlicher Liefertermin 30-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The Quantikine Human Clusterin Immunoassay is a 4.5 hour solid-phase ELISA designed to measure human Clusterin in cell culture supernates, serum, plasma, urine, and saliva. It contains NS0-expressed recombinant human Clusterin and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human Clusterin showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring human Clusterin.
Spezifikation

Spezifikation

3.12 - 200ng/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
1191
6 Plates
1.05ng/mL
Cell Culture Supernates (50μL), Serum (10μL), EDTA Plasma (10μL), Heparin Plasma (10μL), Saliva (25μL), Urine (13μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Serum (10uL), EDTA Plasma (10uL), Heparin Plasma (10uL), Saliva (25uL), Urine (13uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos