Learn More
Human DNAJC13 (RME-8) knockout HeLa cell lysate, 1 Kit
Product Code. 32157949 Shop All Abcam Products
Product Code. 32157949 Supplier Abcam Supplier No. ab2588471Kit
 Restricted Item
Verfügbar ab: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human DNAJC13 (RME-8) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: DNAJC13 (RME-8) knockout HeLa