Learn More
Invitrogen™ Human EARS2 (aa 345-434) Control Fragment Recombinant Protein
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60312 (PA5-60312. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Zugriffsnummer | Q5JPH6 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 124454 |
| Name | Human EARS2 (aa 345-434) Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | 3230401I01Rik; COXPD12; Ears2; GluRS; glutamate tRNA ligase 2, mitochondrial; glutamate--tRNA ligase; Glutamyl-tRNA synthetase; glutamyl-tRNA synthetase 2 (mitochondrial)(putative); glutamyl-tRNA synthetase 2 mitochondrial (putative); glutamyl-tRNA synthetase 2, mitochondrial; Kiaa1970; mKIAA1970; MSE1; Probable glutamate--tRNA ligase, mitochondrial; probable glutamyl-tRNA synthetase, mitochondrial; RGD1307904 |
| Gebräuchliche Bezeichnung | EARS2 |
| Mehr anzeigen |