Learn More
ACROBiosystems Human EMMPRIN / CD147 Protein, Fc Tag
314.52 € - 1959.64 €
Spezifikation
Zugriffsnummer | NP_940991.1 |
---|---|
Molekulargewicht | 46.3 kDa |
Quelle | Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1). |
Immunogen | Ala 22 - His 205 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15886267
|
ACROBiosystems
CD7-H5259-1MG |
1 mg |
1959.64 €
1 Milligramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15876267
|
ACROBiosystems
CD7-H5259-100UG |
100 μg |
314.52 €
100 Mikrogramm |
Verfügbar ab: 26-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
CD147 is also known as Basigin (BSG), or extracellular matrix metalloproteinase inducer (EMMPRIN). The human basigin protein contains 269 amino acids that form two heavily glycosylated C2 type immunoglobulin-like domains at the N-terminal extracellular portion. A second form of basigin has also been characterized that contains one additional immunoglobulin-like domain in its extracellular portion. As members of the immunoglobulin superfamily play fundamental roles in intercellular recognition involved in various immunologic phenomena, differentiation, and development, basigin is thought also to play a role in intercellular recognition and regulate several distinct functions, such as spermatogenesis, expression of the monocarboxylate transporter and the responsiveness of lymphocytes. Basigin is a type I integral membrane receptor that has many ligands, including the cyclophilin (CyP) proteins Cyp-A and CyP-B and certain integrins.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.3 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_940991.1 | |
Human EMMPRIN, Fc Tag (CD7-H5259) is expressed from human 293 cells (HEK293). It contains AA Ala 22 - His 205 (Accession # NP_940991.1). | |
-20°C | |
EMMPRIN | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
46.3 kDa | |
Ala 22 - His 205 | |
Basigin,BSG,5F7,CD147,EMMPRIN,M6,OK,TCSF | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |